EMAIL THIS PAGE TO A FRIEND
65543 Sigma-Aldrich

Methanol

puriss. p.a., absolute, ACS reagent, ≥99.8% (GC)

Synonym: Methyl alcohol

  • CAS Number 67-56-1

  • Linear Formula CH3OH

  • Molecular Weight 32.04

  •  Beilstein Registry Number 1098229

  •  EC Number 200-659-6

  •  MDL number MFCD00004595

Purchase

Properties

Related Categories Methanol, Solvent by Type, Solvents More...
grade   ACS reagent
vapor density   1.11 (vs air)
vapor pressure   410 mmHg ( 50 °C)
  97.68 mmHg ( 20 °C)
InChI Key   OKKJLVBELUTLKV-UHFFFAOYSA-N
grade   absolute
  puriss. p.a.
assay   ≥99.8% (GC)
autoignition temp.   725 °F
expl. lim.   36 %
impurities   KMnO4-reducing substances, in accordance
  acetone, none detected
  substances darkened by H2SO4, in accordance
  ≤0.0001% free alkali (as NH3)
  ≤0.001% aldehyde (as CH3CHO)
  ≤0.001% ketone (as CH3COCH3)
  ≤0.0025% free acid (as HCOOH)
  ≤0.05% water
  <0.001% formaldehyde (HCHO)
evapn. residue   ≤0.001%
refractive index   n20/D 1.329(lit.)
  n20/D 1.329
bp   64.7 °C(lit.)
mp   −98 °C(lit.)
density   0.791 g/mL at 20 °C
  0.791 g/mL at 25 °C(lit.)
cation traces   Al: ≤0.5 mg/kg
  Ba: ≤0.1 mg/kg
  Bi: ≤0.1 mg/kg
  Ca: ≤0.5 mg/kg
  Cd: ≤0.05 mg/kg
  Co: ≤0.05 mg/kg
  Cr: ≤0.02 mg/kg
  Cu: ≤0.02 mg/kg
  Fe: ≤0.5 mg/kg
  K: ≤0.5 mg/kg
  Li: ≤0.1 mg/kg
  Mg: ≤0.1 mg/kg
  Mn: ≤0.02 mg/kg
  Mo: ≤0.1 mg/kg
  Na: ≤0.5 mg/kg
  Ni: ≤0.1 mg/kg
  Pb: ≤0.1 mg/kg
  Sr: ≤0.1 mg/kg
  Zn: ≤0.1 mg/kg
suitability   in accordance for miscibility

Description

Other Notes

Sales restrictions may apply

A -D suffix exists for administrative purposes only.
All -D packages are 100% the same product, same quality, same specification as the package sizes previously sold without a -D.

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN1230 - class 3 - PG 2 - Methanol
WGK Germany 
1
RTECS 
PC1400000
Flash Point(F) 
48.2 °F
Flash Point(C) 
9 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?