EMAIL THIS PAGE TO A FRIEND
65572 Sigma-Aldrich

Methylamine

purum, ≥99.0%

Synonym: Monomethylamine

Purchase

Properties

Related Categories Chemical Synthesis, Specialty Gases, Synthetic Reagents More...
vapor density   1.08 (20 °C, vs air)
vapor pressure   27 psi ( 20 °C)
InChI Key   BAVYZALUXZFZLV-UHFFFAOYSA-N
grade   purum
assay   ≥99.0%
autoignition temp.   806 °F
expl. lim.   4.9-20.8 %
impurities   ≤0.05% ammonia
  ≤0.05% trimethylamine
  ≤0.2% dimethylamine
bp   -6.3 °C(lit.)
mp   -93 °C(lit.)
density   0.70 g/mL at 20 °C(lit.)

Description

Other Notes

Sales restrictions may apply

Same product — same quality — now Sigma-Aldrich brand

Packaging

Cylinder with net ~1.1 kg

Price and Availability

Safety & Documentation

Safety Information

Signal word 
Danger
Hazard statements 
RIDADR 
UN1061 - class 2 - Methylamine, anhydrous
WGK Germany 
2
RTECS 
PF6300000
Flash Point(F) 
<-22 °F
Flash Point(C) 
<-30 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

control valve

Product #

Description

Add to Cart

99112 Outlet valve No 70688, stainless steel, pkg of 1 ea

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?