• USA Home
  • 65600 - Methylamine hydrochloride

EMAIL THIS PAGE TO A FRIEND
65600 Sigma-Aldrich

Methylamine hydrochloride

purum, ≥98.0% (AT)

  • CAS Number 593-51-1

  • Linear Formula CH3NH2 · HCl

  • Molecular Weight 67.52

  •  Beilstein Registry Number 3588822

  •  EC Number 209-795-0

  •  MDL number MFCD00012849

  •  PubChem Substance ID 329760088

Purchase

Properties

Related Categories Amine Salts, Building Blocks, Chemical Synthesis, Nitrogen Compounds, Organic Building Blocks More...
InChI Key   NQMRYBIKMRVZLB-UHFFFAOYSA-N
grade   purum
assay   ≥98.0% (AT)
bp   225-230 °C/15 mmHg(lit.)
mp   229-233 °C
  231-233 °C(lit.)
solubility   H2O: 1 g + 10 mL clear, colorless

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
RTECS 
PA0603000
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

related product

Product #

Description

Add to Cart

Z567671 Q-Tube pressure tube reactor starter kit, capacity 12 mL
Z567736 Q-Tube pressure tube reactor starter kit, capacity 35 mL

replaced by

Product #

Description

Add to Cart

M0505 Methylamine hydrochloride

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?