• USA Home
  • 65780 - 3-Methyl-p-anisaldehyde

EMAIL THIS PAGE TO A FRIEND
65780 Sigma-Aldrich

3-Methyl-p-anisaldehyde

purum, ≥98.0% (GC)

Synonym: 4-Methoxy-3-methylbenzaldehyde

  • CAS Number 32723-67-4

  • Empirical Formula (Hill Notation) C9H10O2

  • Molecular Weight 150.17

  •  Beilstein Registry Number 2084045

  •  EC Number 251-177-8

  •  MDL number MFCD00003375

  •  PubChem Substance ID 329760107

Purchase

Properties

Related Categories Aldehydes, Building Blocks, C9, Carbonyl Compounds, Chemical Synthesis,
InChI Key   MYLBIQHZWFWSMH-UHFFFAOYSA-N
grade   purum
assay   ≥98.0% (GC)
refractive index   n20/D 1.569
density   1.098 g/mL at 20 °C(lit.)

Price and Availability

Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
Flash Point(F) 
233.6 °F
Flash Point(C) 
112 °C

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?