• USA Home
  • 65877 - 4-(6-Methyl-2-benzothiazolyl)phenyl isocyanate

EMAIL THIS PAGE TO A FRIEND
65877 Sigma-Aldrich

4-(6-Methyl-2-benzothiazolyl)phenyl isocyanate

suitable for fluorescence, ≥98.0% (HPLC)

  • CAS Number 67229-93-0

  • Empirical Formula (Hill Notation) C15H10N2OS

  • Molecular Weight 266.32

  •  Beilstein Registry Number 229420

  •  MDL number MFCD00467423

  •  PubChem Substance ID 24884377

Purchase

Properties

Related Categories Alphabetical, Analytical Reagents, Analytical/Chromatography, Biochemicals and Reagents, Carbohydrate Labels,
InChI Key   DKMGYFQSCKPSEQ-UHFFFAOYSA-N
assay   ≥98.0% (HPLC)
mp   140-150 °C (dec.)
fluorescence   λex 327 nm; λem 382 nm in methylene chloride (after derivatization with hexylamine)
  λex 350 nm; λem 374 nm in chlorobenzene
suitability   suitable for fluorescence
storage temp.   2-8°C

Description

Other Notes

Reagent for HPLC precolumn derivatization of prim., sec. and tert. hydroxy compounds and prim. and sec. amines for fluorimetric determination

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Application

4-(6-Methyl-2-benzothiazolyl)phenyl isocyanate is a fluorogenic agent use to derivatize primary, secondary and tertiary hydroxyl compounds and primary and secondary amines for analytical analysis and chromatographic separation.

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?