EMAIL THIS PAGE TO A FRIEND
65890 Sigma-Aldrich

4-Methylbenzylamine

purum, ≥97.0% (GC)

Synonym: p-Xylylamine

  • CAS Number 104-84-7

  • Linear Formula CH3C6H4CH2NH2

  • Molecular Weight 121.18

  •  Beilstein Registry Number 956670

  •  EC Number 203-243-2

  •  MDL number MFCD00008123

  •  PubChem Substance ID 329760130

Purchase

Properties

InChI Key   HMTSWYPNXFHGEP-UHFFFAOYSA-N
grade   purum
assay   ≥97.0% (GC)
refractive index   n20/D 1.534(lit.)
  n20/D 1.536
bp   195 °C(lit.)
mp   12-13 °C(lit.)
transition temp   solidification point 12-15 °C(lit.)
density   0.952 g/mL at 25 °C(lit.)

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS05  GHS05
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 2735PSN1 8 / PGII
WGK Germany 
3
Flash Point(F) 
176 °F
Flash Point(C) 
80 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

M31201 4-Methylbenzylamine, 97%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?