EMAIL THIS PAGE TO A FRIEND
66020 Sigma-Aldrich

3-Methyl-2-butanol

purum, ≥98.0% (GC)

Synonym: Isopropyl methyl carbinol

  • CAS Number 598-75-4

  • Linear Formula (CH3)2CHCH(OH)CH3

  • Molecular Weight 88.15

  •  Beilstein Registry Number 1718800

  •  EC Number 209-950-2

  •  MDL number MFCD00004527

  •  PubChem Substance ID 24884464

Purchase

Properties

Related Categories Alcohols, Building Blocks, C2 to C6, Chemical Synthesis, Organic Building Blocks,
InChI Key   MXLMTQWGSQIYOW-UHFFFAOYSA-N
grade   purum
assay   ≥98.0% (GC)
refractive index   n20/D 1.409(lit.)
  n20/D 1.409
bp   112 °C(lit.)
density   0.818 g/mL at 25 °C(lit.)

Description

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Warning
Hazard statements 
Precautionary statements 
Supplemental Hazard Statements 
Repeated exposure may cause skin dryness or cracking.
RIDADR 
UN1105 - class 3 - PG 3 - Pentanols
WGK Germany 
1
Flash Point(F) 
93.2 °F
Flash Point(C) 
34 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?