• USA Home
  • 66095 - 3-Methyl-3-buten-1-ol

EMAIL THIS PAGE TO A FRIEND
66095 Sigma-Aldrich

3-Methyl-3-buten-1-ol

purum, ≥98.0% (GC)

  • CAS Number 763-32-6

  • Linear Formula CH2=C(CH3)CH2CH2OH

  • Molecular Weight 86.13

  •  Beilstein Registry Number 1071239

  •  EC Number 212-110-8

  •  MDL number MFCD00002933

  •  PubChem Substance ID 329760184

Purchase

Properties

Related Categories Acyclic, Alkenes, Building Blocks, Chemical Synthesis, Organic Building Blocks More...
InChI Key   CPJRRXSHAYUTGL-UHFFFAOYSA-N
grade   purum
assay   ≥98.0% (GC)
refractive index   n20/D 1.433(lit.)
  n20/D 1.434
bp   130-132 °C(lit.)
density   0.853 g/mL at 25 °C(lit.)

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 1987C 3 / PGIII
WGK Germany 
1
Flash Point(F) 
107.6 °F
Flash Point(C) 
42 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

129402 3-Methyl-3-buten-1-ol, 97%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?