• USA Home
  • 66120 - 2-Methyl-3-butyn-2-ol

EMAIL THIS PAGE TO A FRIEND
66120 Sigma-Aldrich

2-Methyl-3-butyn-2-ol

purum, ≥99.0% (GC)

Synonym: Dimethyl ethynyl carbinol

  • CAS Number 115-19-5

  • Linear Formula HC≡CC(CH3)2OH

  • Molecular Weight 84.12

  •  Beilstein Registry Number 635746

  •  EC Number 204-070-5

  •  MDL number MFCD00004467

Purchase

Properties

Related Categories Alkynes, Building Blocks, Chemical Synthesis, Organic Building Blocks, Terminal More...
vapor pressure   15 mmHg ( 20 °C)
InChI Key   CEBKHWWANWSNTI-UHFFFAOYSA-N
grade   purum
assay   ≥99.0% (GC)
autoignition temp.   662 °F
expl. lim.   16.6 %
refractive index   n20/D 1.42(lit.)
  n20/D 1.420
bp   104 °C(lit.)
mp   2.6 °C(lit.)
density   0.868 g/mL at 25 °C(lit.)

Description

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 1987BF 3 / PGII
WGK Germany 
1
Flash Point(F) 
<69.8 °F
Flash Point(C) 
<21 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

replaced by

Product #

Description

Add to Cart

129763 2-Methyl-3-butyn-2-ol, 98%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?