EMAIL THIS PAGE TO A FRIEND
66295 Sigma-Aldrich

Methylcyclohexane

purum, absolute, over molecular sieve (H2O ≤0.005%), ≥98.0% (GC)

Synonym: Hexahydrotoluene

Purchase

Properties

Related Categories Anhydrous Solvents, Solvent by Application, Solvents More...
vapor density   3.4 (vs air)
vapor pressure   37 mmHg ( 20 °C)
  83.2 mmHg ( 37.7 °C)
InChI Key   UAEPNZWRGJTJPN-UHFFFAOYSA-N
grade   absolute
  purum
assay   ≥98.0% (GC)
autoignition temp.   545 °F
quality   over molecular sieve (H2O ≤0.005%)
expl. lim.   6.7 %
refractive index   n20/D 1.422(lit.)
  n20/D 1.423
bp   101 °C(lit.)
mp   −126 °C(lit.)
density   0.77 g/mL at 25 °C(lit.)

Price and Availability

Safety & Documentation

Safety Information

Signal word 
Danger
Hazard statements 
RIDADR 
UN 2296 3 / PGII
WGK Germany 
2
RTECS 
GV6125000
Flash Point(F) 
24.8 °F
Flash Point(C) 
-4 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?