EMAIL THIS PAGE TO A FRIEND
66315 Sigma-Aldrich

Peptone Test Kit

for microbiology

Synonym: Test kit of different peptones

Purchase

Properties

Related Categories Identification Tests & Reagents, Microbiology, Test Kits More...
grade   for microbiology

Description

Components

22090 Casein Hydrolysate
61300 Lactalbumin Hydrolysate
61302 Lactalbumin Hydrolysate
82303 Peptone enzymatic digest from Casein
70951 Peptone enzymatic digest from Gelatine
87972 Peptone enzymatic digest from soybean
77180 Peptone from animal proteins
70173 Peptone from casein and other animal proteins
70171 Peptone from casein, acid digest
70169 Peptone from casein, pancreatic digest
70172 Peptone from casein, tryptic digest
70176 Peptone from gelatine, pancreatic digest
70177 Peptone from lactalbumin, enzymatic digest, readily soluble
70175 Peptone from meat, enzymatic digest
82962 Peptone from meat, enzymatic digest
70174 Peptone from meat, peptic digest
83059 Peptone from potatoes
70178 Peptone from soybean meal, enzymatic digest
S1674 Soy protein acid hydrolysate
82514 Protein Hydrolysate Amicase
82450 Proteose-Peptone
95039 Tryptone enzymatic digest from Casein
93733 Tryptose

Application

This kit helps one to find the best peptide source for a microorganism. Test a choice of 23 peptones and hydrolysates for a fermentation, a growth, or for a diagnostic medium. With a peptide source specifically fitted to a microorganism, it is possible to increase the yield and improve the reproducibility of your results.

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles

Articles

EZ-Product Family for Bioburden Testing

The EZ family of products has been designed to optimize and streamline the bioburden workflow. The family includes:
Keywords: Filtration, Separation

Microbiology Introduction

An initial aim of all microbiologists is the reproducible growth of their microbial cultures, no matter whether the microorganisms are of natural origin or have been genetically engineered by man. Re...
Keywords: Absorption, Aerobic, Anaerobic, Antibiotics, Antimicrobials, Buffers, Culture media, Diffusion, Environmental, Filtration, Genetic, Growth factors, Metabolism, Phase transitions, Precipitation, Reductions, Respiration, Sample preparations, Vitamins

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?