EMAIL THIS PAGE TO A FRIEND
66360 Sigma-Aldrich

4-Methylcyclohexanol

purum, cis+trans, ≥98.0% (GC)

Synonym: Hexahydro-p-cresol

  • CAS Number 589-91-3

  • Empirical Formula (Hill Notation) C7H14O

  • Molecular Weight 114.19

  •  Beilstein Registry Number 1445998

  •  EC Number 209-664-8

  •  MDL number MFCD00001449

  •  PubChem Substance ID 329760244

Purchase

Properties

Related Categories Alcohols, Building Blocks, C7 to C8, Chemical Synthesis, Organic Building Blocks,
InChI Key   MQWCXKGKQLNYQG-UHFFFAOYSA-N
grade   purum
assay   ≥98.0% (GC)
refractive index   n20/D 1.459
bp   170-175 °C(lit.)
density   0.916 g/mL at 20 °C(lit.)

Price and Availability

Safety & Documentation

Safety Information

WGK Germany 
3
Flash Point(F) 
158 °F
Flash Point(C) 
70 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

153095 4-Methylcyclohexanol, mixture of cis and trans, 98%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?