• USA Home
  • 66380 - 2-Methylcyclohexanone

EMAIL THIS PAGE TO A FRIEND
66380 Sigma-Aldrich

2-Methylcyclohexanone

purum, ≥98.0% (GC)

Synonym: Tetrahydro-o-cresol

  • CAS Number 583-60-8

  • Linear Formula CH3C6H9(=O)

  • Molecular Weight 112.17

  •  Beilstein Registry Number 506751

  •  EC Number 209-513-6

  •  MDL number MFCD00001635

  •  PubChem Substance ID 24884659

Purchase

Properties

Related Categories Building Blocks, C7 to C8, Carbonyl Compounds, Chemical Synthesis, Ketones,
InChI Key   LFSAPCRASZRSKS-UHFFFAOYSA-N
grade   purum
assay   ≥98.0% (GC)
refractive index   n20/D 1.448(lit.)
  n20/D 1.448
bp   162-163 °C(lit.)
density   0.924 g/mL at 25 °C(lit.)

Description

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
UN 2297 3 / PGIII
WGK Germany 
1
RTECS 
GW1750000
Flash Point(F) 
118.4 °F
Flash Point(C) 
48 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?