• USA Home
  • 66460 - N-Methylcyclohexylamine

EMAIL THIS PAGE TO A FRIEND
66460 Sigma-Aldrich

N-Methylcyclohexylamine

purum, ≥98.0% (GC)

Synonym: N-Cyclohexylmethylamine

Purchase

Properties

vapor density   4 (vs air)
vapor pressure   11 mmHg ( 40 °C)
InChI Key   XTUVJUMINZSXGF-UHFFFAOYSA-N
grade   purum
assay   ≥98.0% (GC)
refractive index   n20/D 1.456(lit.)
  n20/D 1.456
bp   149 °C(lit.)
mp   −9-−7 °C(lit.)
density   0.868 g/mL at 25 °C(lit.)

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 2734PSN1 3(8) / PGII
WGK Germany 
1
RTECS 
GX1529000
Flash Point(F) 
84.2 °F
Flash Point(C) 
29 °C
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?