• USA Home
  • 66674 - Levan from Zymomonas mobilis

EMAIL THIS PAGE TO A FRIEND
66674 Sigma-Aldrich

Levan from Zymomonas mobilis

Purchase

Description

Application

Levan, a homopolysaccharide composed of β-(2-6)-D-fructofuranosyl, is related to inulin (β-(2-1)-fructofuranose). Levan possesses anti-inflammatory and cell proliferative properties that make it useful in the development of cosmeceuticals. Levan is used in the food industry as a thickening agent and source of fructose.

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?