• USA Home
  • 66715 - 4-Amino-2-chloro-6,7-dimethoxyquinazoline

EMAIL THIS PAGE TO A FRIEND
66715 Sigma-Aldrich

4-Amino-2-chloro-6,7-dimethoxyquinazoline

puriss., ≥99.0% (HPLC)

Synonym: 2-Chloro-6,7-dimethoxy-4-quinazolinamine

  • CAS Number 23680-84-4

  • Empirical Formula (Hill Notation) C10H10ClN3O2

  • Molecular Weight 239.66

  •  Beilstein Registry Number 747463

  •  EC Number 245-821-7

  •  MDL number MFCD00051734

  •  PubChem Substance ID 329760308

Purchase

Properties

Related Categories Building Blocks, Chemical Synthesis, Halogenated Heterocycles, Heterocyclic Building Blocks, Quinazolines More...
InChI Key   HWIIAAVGRHKSOJ-UHFFFAOYSA-N
grade   puriss.
assay   ≥99.0% (HPLC)
mp   262-268 °C (dec.)(lit.)

Description

Other Notes

Building block for preparing terazosine and its analogues, α1-adrenoceptor antagonists, see e.g.

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

518654 4-Amino-2-chloro-6,7-dimethoxyquinazoline, 95%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?