• USA Home
  • 66916 - Methyl α-D-galactopyranoside

EMAIL THIS PAGE TO A FRIEND
66916 Sigma-Aldrich

Methyl α-D-galactopyranoside

purum, ≥98.0% (sum of enantiomers, HPLC)

Synonym: Methyl α-D-galactoside

  • CAS Number 3396-99-4

  • Empirical Formula (Hill Notation) C7H14O6

  • Molecular Weight 194.18

  •  Beilstein Registry Number 81570

  •  EC Number 222-251-7

  •  MDL number MFCD00064085

  •  PubChem Substance ID 24885022

Purchase

Properties

Related Categories Carbohydrate Chemistry, Carbohydrate Derivatives, Chemical Biology, Chemical Synthesis
InChI Key   HOVAGTYPODGVJG-PZRMXXKTSA-N
grade   purum
assay   ≥98.0% (sum of enantiomers, HPLC)
optical activity   [α]20/D +169±3°, c = 1.5% in methanol
impurities   ~8.5% water
mp   116-117 °C(lit.)

Description

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

replaced by

Product #

Description

Add to Cart

M1379 Methyl α-D-galactopyranoside

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?