• USA Home
  • 66990 - 3-Methylglutaric acid

EMAIL THIS PAGE TO A FRIEND
66990 Sigma-Aldrich

3-Methylglutaric acid

purum, ≥98.0% (GC)

  • CAS Number 626-51-7

  • Linear Formula CH3CH(CH2COOH)2

  • Molecular Weight 146.14

  •  Beilstein Registry Number 1759502

  •  EC Number 210-951-5

  •  MDL number MFCD00002727

  •  PubChem Substance ID 329760359

Purchase

Properties

Related Categories Building Blocks, C6, Carbonyl Compounds, Carboxylic Acids, Chemical Synthesis,
InChI Key   XJMMNTGIMDZPMU-UHFFFAOYSA-N
grade   purum
assay   ≥98.0% (GC)
mp   81-86 °C(lit.)
  81-86 °C

Price and Availability


Laboratory Gloves
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?