EMAIL THIS PAGE TO A FRIEND
67076 Sigma-Aldrich

1-Methyluric acid

≥98.0% (HPLC)

Synonym: 1-Methyl-2,6,8-trihydroxypurine, 1-U, 1MUA

  • CAS Number 708-79-2

  • Empirical Formula (Hill Notation) C6H6N4O3

  • Molecular Weight 182.14

  •  Beilstein Registry Number 195190

  •  EC Number 211-905-7

  •  MDL number MFCD00042987

  •  PubChem Substance ID 329760385

Purchase

Properties

Related Categories Building Blocks, Chemical Synthesis, Heterocyclic Building Blocks, Purines
InChI Key   QFDRTQONISXGJA-UHFFFAOYSA-N
assay   ≥98.0% (HPLC)

Description

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles

Articles

HPLC Analysis of Caffeine Metabolites and Analogs on Discovery® C18

From our library of Articles, Sigma-Aldrich presents HPLC Analysis of Caffeine Metabolites and Analogs on Discovery® C18
Keywords: Chromatography, Clinical, High performance liquid chromatography

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?