673781 Sigma-Aldrich

Acetone

ACS reagent, ≥99.5%

DOWNLOAD MSDS (PDF)

Properties

Related Categories Aluminum Bottles, Solvent Bottles, Solvent Packaging Options, Solvents
grade   ACS reagent
vapor density   2 (vs air)
vapor pressure   184 mmHg ( 20 °C)
assay   ≥99.5%
expl. lim.   13.2 %
impurities   ≤0.0003 meq/g Titr. acid
  ≤0.0006 meq/g Titr. base
  ≤0.002% aldehyde as formaldehyde
  ≤0.05% isopropanol
  ≤0.05% methanol
  ≤0.5% water
evapn. residue   ≤0.001%
color   APHA: ≤10
refractive index   n20/D 1.359(lit.)
bp   56 °C/760 mmHg(lit.)
mp   −94 °C(lit.)
density   0.791 g/mL at 25 °C(lit.)

Description

Packaging

1, 6×1, 4, 4×4 L in aluminum bottle

200 L in steel drum

Application

Acetone′s luminesence intensity is dependent upon the solution components 1. The absorption of UV light by acetone, results in its photolysis and the production of radials 1.

Price and Availability

Customers Also Viewed

Acetone
650501Acetone

CHROMASOLV® Plus, for HPLC, ≥99.9%

Acetone
154598Acetone

ACS spectrophotometric grade, ≥99.5%

Acetone
179124Acetone

ACS reagent, ≥99.5%

Acetone
320110Acetone

ACS reagent, ≥99.5%

Acetone
179973Acetone

Laboratory Reagent, ≥99.5%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?