673811 Sigma-Aldrich

Diethyl ether

anhydrous, ACS reagent, ≥99.0%, contains BHT as inhibitor

DOWNLOAD MSDS (PDF)

Synonym: Ether, Ethyl ether

Properties

Related Categories Aluminum Bottles, Diethyl Ether, Solvent by Type, Solvent Bottles, Solvent Packaging Options,
grade   ACS reagent
  anhydrous
vapor density   2.6 (vs air)
vapor pressure   28.66 psi ( 55 °C)
  8.56 psi ( 20 °C)
assay   ≥99.0%
autoignition temp.   320 °F
contains   BHT as inhibitor
expl. lim.   36.5 %
impurities   Alcohol (as CH3CH2OH), passes test
  ≤0.0002 meq/g Titr. acid
  ≤0.001% Carbonyl (as HCHO)
  ≤0.03% Water
  ≤1 ppm Peroxide (as H2O2)
evapn. residue   ≤0.001%
color   APHA: ≤10
refractive index   n20/D 1.3530(lit.)
bp   34.6 °C(lit.)
mp   −116 °C(lit.)
density   0.706 g/mL at 25 °C(lit.)

Description

Packaging

1, 6×1, 4, 4×4 L in aluminum bottle

20 L in steel drum

200 L in Pure-Pac™ 1

200 L in steel drum

Price and Availability

Customers Also Viewed

Diethyl ether

contains 1 ppm BHT as inhibitor, anhydrous, ≥99.7%

Diethyl ether

CHROMASOLV®, for HPLC, ≥99.9%, inhibitor-free

Diethyl ether

ACS reagent, anhydrous, ≥99.0%, contains BHT as inhibitor

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?