• USA Home
  • 67456 - Fluorous Biphase Catalysis, Kit 1: C-C Coupling

EMAIL THIS PAGE TO A FRIEND
67456 Sigma-Aldrich

Fluorous Biphase Catalysis, Kit 1: C-C Coupling

Purchase

Description

Components

95447 Bis[tris(4-(1H,1H,2H,2H-perfluorodecyl)phenyl)phosphine]palladium(II) dichloride, 100 mg;
78181 Phenylboronic acid, 1 g;
56647 Tributyl-(2-furyl)stannane, 1 mL;
53482 Methyl 4-bromobenzoate, 2.5 g
73036 Lithium chloride anhydrous, 5 g;
40248 N,N-Dimethylformamide, 250 mL;
77280 (Trifluoromethyl)undecafluorocyclohexane 25 mL;
38568 1,2-Dimethoxyethane 50 mL;
60239 Potassium fluoride 5 g

Application

Contains the components necessary to perform up to 10 C-C coupling reactions under fluorous biphasic conditions including 2 model reactions (Stille and Suzuki coupling) to familiarize with Fluorous Phase Organic Synthesis. Analytical methods and reference spectra are given to reconfirm the results of the 2 model reactions.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
UN 1812 6.1/PG 2

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?