EMAIL THIS PAGE TO A FRIEND
67592 Sigma-Aldrich

Boldine

analytical reference material

Synonym: 2,9-Dihydroxy-1,10-dimethoxyaporphine

  • CAS Number 476-70-0

  • Empirical Formula (Hill Notation) C19H21NO4

  • Molecular Weight 327.37

  •  Beilstein Registry Number 94036

  •  EC Number 207-509-9

  •  MDL number MFCD00135040

  •  PubChem Substance ID 329760629

Purchase

Properties

Related Categories A - D, Alphabetical Index of Phytochemical Standards, Analytical Standards, Analytical/Chromatography, Peumus,
grade   analytical reference material
InChI Key   LZJRNLRASBVRRX-ZDUSSCGKSA-N
assay   ≥96.5% (qNMR)
  ≥97.5% (HPLC)
form   neat
shelf life   limited shelf life, expiry date on the label
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
mp   157-164 °C
format   neat

Description

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
RTECS 
CE0750000
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?