EMAIL THIS PAGE TO A FRIEND
67600 Sigma-Aldrich

2-Methylindole

purum, ≥97.5% (HPLC)

Synonym: NSC 7514

  • CAS Number 95-20-5

  • Empirical Formula (Hill Notation) C9H9N

  • Molecular Weight 131.17

  •  Beilstein Registry Number 109781

  •  EC Number 202-398-3

  •  MDL number MFCD00005616

  •  PubChem Substance ID 329760631

Purchase

Properties

Related Categories Building Blocks, C7 to C9, Chemical Synthesis, Heterocyclic Building Blocks, Indoles More...
InChI Key   BHNHHSOHWZKFOX-UHFFFAOYSA-N
grade   purum
assay   ≥97.5% (HPLC)
bp   273 °C(lit.)
mp   56-60 °C
  57-59 °C(lit.)

Description

Caution

may discolor on storage

Application

Reactant for:
• Regioselective synthesis of oxopyrrolidine analogs via iodine-catalyzed Markovnikov addition reaction
• Friedel-Crafts alkylation reactions
• Preparation of tryptophan dioxygenase inhibitors pyridyl-ethenyl-indoles as potential anticancer immunomodulators
• Preparation of plant-growth inhibitors
• Michael addition reactions
• Synthesis of cyclooxygenase-1 (COX-1)/cyclooxygenase-2 (COX-2) inhibitors

Price and Availability


Laboratory Gloves
Safety & Documentation

Safety Information

Symbol 
GHS05  GHS05
Signal word 
Danger
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
RTECS 
NM0345000
Flash Point(F) 
285.8 °F
Flash Point(C) 
141 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

M51407 2-Methylindole, 98%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?