• USA Home
  • 67760 - Diethyl methylmalonate

EMAIL THIS PAGE TO A FRIEND
67760 Sigma-Aldrich

Diethyl methylmalonate

puriss., ≥99.0% (GC)

Synonym: Methylmalonic acid diethyl ester

  • CAS Number 609-08-5

  • Linear Formula CH3CH(COOC2H5)2

  • Molecular Weight 174.19

  •  Beilstein Registry Number 637028

  •  EC Number 210-175-7

  •  MDL number MFCD00009162

  •  PubChem Substance ID 329760484

Purchase

Properties

Related Categories Building Blocks, C8 to C9, Carbonyl Compounds, Chemical Synthesis, Esters,
InChI Key   UPQZOUHVTJNGFK-UHFFFAOYSA-N
grade   puriss.
assay   ≥99.0% (GC)
refractive index   n20/D 1.413(lit.)
  n20/D 1.414
bp   198-199 °C(lit.)
  78-80 °C/10 mmHg
density   1.022 g/mL at 20 °C(lit.)

Price and Availability

Safety & Documentation

Safety Information

WGK Germany 
3
Flash Point(F) 
179.6 °F
Flash Point(C) 
82 °C
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?