• USA Home
  • 67849 - tert-Amyl methyl ether

EMAIL THIS PAGE TO A FRIEND
67849 Sigma-Aldrich

tert-Amyl methyl ether

purum, absolute, over molecular sieve (H2O ≤0.005%), ≥97.0% (GC)

Synonym: 1,1-Dimethylpropyl methyl ether, Methyl 2-methyl-2-butyl ether, Methyl tert-pentyl ether, TAME

  • CAS Number 994-05-8

  • Linear Formula C2H5C(CH3)2OCH3

  • Molecular Weight 102.17

  •  Beilstein Registry Number 1731452

  •  EC Number 213-611-4

  •  MDL number MFCD00010186

  •  PubChem Substance ID 329760696

Purchase

Properties

InChI Key   HVZJRWJGKQPSFL-UHFFFAOYSA-N
grade   absolute
  purum
assay   ≥97.0% (GC)
quality   over molecular sieve (H2O ≤0.005%)
refractive index   n20/D 1.389(lit.)
  n20/D 1.389
  n20/D 1.3896
bp   85-86 °C(lit.)
density   0.76-0.78 g/mL at 25 °C(lit.)

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 3271 3 / PGII
WGK Germany 
3
RTECS 
EK4421000
Flash Point(F) 
19.4 °F
Flash Point(C) 
-7 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?