• USA Home
  • 67874 - 4-Methylmorpholine N-oxide solution

EMAIL THIS PAGE TO A FRIEND
67874 Sigma-Aldrich

4-Methylmorpholine N-oxide solution

technical, ~50% in H2O

Synonym: NMMO solution, NMO solution, NSC 73198, NSC 82153

  • CAS Number 7529-22-8

  • Empirical Formula (Hill Notation) C5H11NO2

  • Molecular Weight 117.15

  •  MDL number MFCD00005947

  •  PubChem Substance ID 24885452

Purchase

Properties

Related Categories Chemical Synthesis, Other Oxidizing Agents, Oxidizing Agents, Synthetic Reagents
InChI Key   LFTLOKWAGJYHHR-UHFFFAOYSA-N
grade   technical
concentration   ~50% in H2O
refractive index   n20/D 1.43

Description

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Application

Oxidant for synthesis of novel organic polymer - inorganic hybrid for use as a catalyst for asymmetrical epoxidation

Reagent or Reactant for:
• Cyclocondensation and cyclization in the enantioselective synthesis of oxazolomycin A
• Wharton rearrangement and stereoselective dihydroxylation reactions
• Reduction of amine N-oxides by diboron reagents
• Synthesis of polysaccharide blend fibers
• Synthesis of cellulose / modified nano-SiO2 composite packaging films
• Copper-catalyzed oxidative coupling for preparation of propargylamines

Used as a pretreatment for techno-economical study of ethanol and biogas production from spruce wood

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
NONH for all modes of transport
WGK Germany 
1

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?