EMAIL THIS PAGE TO A FRIEND
67990 Sigma-Aldrich

1-Acetonaphthone

purum, ≥96.0% (GC)

Synonym: 1-Acetylnaphthalene, Methyl 1-naphthyl ketone

Purchase

Properties

Related Categories Building Blocks, C11 to C12, Carbonyl Compounds, Chemical Synthesis, Ketones,
InChI Key   QQLIGMASAVJVON-UHFFFAOYSA-N
grade   purum
assay   ≥96.0% (GC)
impurities   ~3% 2-acetonaphthone
refractive index   n20/D 1.628(lit.)
  n20/D 1.628
bp   302 °C(lit.)
mp   10.5 °C(lit.)
density   1.12 g/mL at 25 °C(lit.)

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
NONH for all modes of transport
WGK Germany 
2
RTECS 
AL2982800
Flash Point(F) 
235.4 °F
Flash Point(C) 
113 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

276766 1-Acetonaphthone, 97%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?