• USA Home
  • 680265 - Mercury(I) nitrate dihydrate

EMAIL THIS PAGE TO A FRIEND
680265 Sigma-Aldrich

Mercury(I) nitrate dihydrate

reagent grade, ≥97%

Synonym: Mercurous nitrate dihydrate

Purchase

Properties

Related Categories Catalysis and Inorganic Chemistry, Chemical Synthesis, Essential Chemicals, Mercury, Mercury Salts,
grade   reagent grade
assay   ≥97%
density   4.78 g/mL at 25 °C(lit.)

Description

Other Notes

A -D suffix exists for administrative purposes only.
All -D packages are 100% the same product, same quality, same specification as the package sizes previously sold without a -D.

Packaging

50, 250 g in glass bottle

Price and Availability

Safety & Documentation

Documents

Certificate of Analysis

Certificate of Origin

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?