• USA Home
  • 68253 - Sodium 3-methyl-2-oxobutyrate

EMAIL THIS PAGE TO A FRIEND
68253 Sigma-Aldrich

Sodium 3-methyl-2-oxobutyrate

purum, ≥97.0% (NT)

Synonym: α-Ketoisovaleric acid sodium salt, 2-Keto-3-methylbutyric acid sodium salt, 3-Methyl-2-oxobutanoic acid sodium salt, 3-Methyl-2-oxobutyric acid sodium salt, Ketovaline sodium salt

  • CAS Number 3715-29-5

  • Linear Formula (CH3)2CHCOCOONa

  • Molecular Weight 138.10

  •  Beilstein Registry Number 4334552

  •  EC Number 223-062-2

  •  MDL number MFCD00002581

  •  PubChem Substance ID 24885721

Purchase

Properties

Related Categories Building Blocks, Carbonyl Compounds, Carboxylic Acid Salts, Chemical Synthesis, Organic Building Blocks More...
InChI Key   WIQBZDCJCRFGKA-UHFFFAOYSA-M
grade   purum
assay   ≥97.0% (NT)
mp   220-230 °C (dec.)(lit.)

Description

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

replaced by

Product #

Description

Add to Cart

198994 Sodium 3-methyl-2-oxobutyrate, 95%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?