EMAIL THIS PAGE TO A FRIEND
68551 Sigma-Aldrich

2-Bromopropane

analytical standard

Synonym: Isopropyl bromide

  • CAS Number 75-26-3

  • Linear Formula (CH3)2CHBr

  • Molecular Weight 122.99

  •  Beilstein Registry Number 741852

  •  EC Number 200-855-1

  •  MDL number MFCD00000147

  •  PubChem Substance ID 24885840

Purchase

Properties

Related Categories A-B, A-C, Additional Standards, Alpha Sort, Analytical Standards,
grade   analytical standard
InChI Key   NAMYKGVDVNBCFQ-UHFFFAOYSA-N
assay   ≥99.5% (GC)
form   neat
shelf life   limited shelf life, expiry date on the label
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
refractive index   n20/D 1.425(lit.)
  n20/D 1.425
bp   59 °C(lit.)
mp   −89 °C(lit.)
density   1.31 g/mL at 25 °C(lit.)
format   neat

Description

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
Supplemental Hazard Statements 
Repeated exposure may cause skin dryness or cracking.
RIDADR 
UN 2344 3 / PGII
WGK Germany 
1
RTECS 
TX4111000
Flash Point(F) 
66.2 °F
Flash Point(C) 
19 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?