• USA Home
  • 68752 - 3-Methyl-1-phenyl-4-trifluoroacetyl-2-pyrazolin-5-one

EMAIL THIS PAGE TO A FRIEND
68752 Sigma-Aldrich

3-Methyl-1-phenyl-4-trifluoroacetyl-2-pyrazolin-5-one

for HPLC derivatization, ≥98.0%

Synonym: 5-Hydroxy-3-methyl-1-phenyl-4-trifluoroacetylpyrazole

  • CAS Number 1691-93-6

  • Empirical Formula (Hill Notation) C12H9F3N2O2

  • Molecular Weight 270.21

  •  Beilstein Registry Number 5969762

  •  MDL number MFCD00143378

  •  PubChem Substance ID 329761135

Purchase

Properties

Related Categories Analytical Reagents, Analytical/Chromatography, Derivatization Reagents, Derivatization Reagents HPLC, UV-VIS More...
grade   for HPLC derivatization
InChI Key   VXIIAPOORVGKBU-UHFFFAOYSA-N
assay   ≥98.0% (T)
  ≥98.0%
mp   145-148 °C

Price and Availability


Laboratory Gloves
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?