EMAIL THIS PAGE TO A FRIEND
68810 Sigma-Aldrich

1-Methylpiperazine

purum, ≥99.0% (GC)

  • CAS Number 109-01-3

  • Empirical Formula (Hill Notation) C5H12N2

  • Molecular Weight 100.16

  •  Beilstein Registry Number 102724

  •  EC Number 203-639-5

  •  MDL number MFCD00005966

  •  PubChem Substance ID 329760834

Purchase

Properties

Related Categories Building Blocks, C4 to C8, Chemical Synthesis, Heterocyclic Building Blocks, Piperazines More...
vapor density   3.5 (vs air)
InChI Key   PVOAHINGSUIXLS-UHFFFAOYSA-N
grade   purum
assay   ≥99.0% (GC)
refractive index   n20/D 1.466(lit.)
  n20/D 1.466
bp   138 °C(lit.)
density   0.903 g/mL at 25 °C(lit.)

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 2734PSN1 3(8) / PGII
WGK Germany 
2
RTECS 
TM1225000
Flash Point(F) 
102.2 °F
Flash Point(C) 
39 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?