EMAIL THIS PAGE TO A FRIEND
68815 Sigma-Aldrich

2-Methylpiperazine

purum, ≥98.0% (GC)

  • CAS Number 109-07-9

  • Empirical Formula (Hill Notation) C5H12N2

  • Molecular Weight 100.16

  •  Beilstein Registry Number 383566

  •  EC Number 203-644-2

  •  MDL number MFCD00005954

  •  PubChem Substance ID 329760842

Purchase

Properties

Related Categories Building Blocks, C4 to C8, Chemical Synthesis, Heterocyclic Building Blocks, Piperazines More...
InChI Key   JOMNTHCQHJPVAZ-UHFFFAOYSA-N
grade   purum
assay   ≥98.0% (GC)
bp   155 °C/763 mmHg(lit.)
mp   61-63 °C(lit.)
  61-63 °C

Price and Availability


Laboratory Gloves
Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN1325 - class 4.1 - PG 2 - Flammable solids, organic, n.o.s., HI: all
WGK Germany 
2
RTECS 
TM1225020
Flash Point(F) 
149 °F
Flash Point(C) 
65 °C
Protocols & Articles
Related Products

replaced by

Product #

Description

Add to Cart

M72404 2-Methylpiperazine, 95%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?