EMAIL THIS PAGE TO A FRIEND
68900 Sigma-Aldrich

N-Methylpropionamide

purum

  • CAS Number 1187-58-2

  • Linear Formula CH3CH2CONHCH3

  • Molecular Weight 87.12

  •  Beilstein Registry Number 1737640

  •  EC Number 214-699-7

  •  MDL number MFCD00009300

Purchase

Properties

Related Categories Amides, Building Blocks, C2 to C7, Carbonyl Compounds, Chemical Synthesis,
InChI Key   QJQAMHYHNCADNR-UHFFFAOYSA-N
grade   purum
refractive index   n20/D 1.436(lit.)
  n20/D 1.437
bp   146 °C/90 mmHg(lit.)
mp   −43 °C(lit.)
density   0.931 g/mL at 25 °C(lit.)

Description

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
RTECS 
UE4515000
Flash Point(F) 
222.8 °F
Flash Point(C) 
106 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

replaced by

Product #

Description

Add to Cart

290874 N-Methylpropionamide, 98%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?