• USA Home
  • 69118 - 1-Methyl-2-pyrrolidinone

EMAIL THIS PAGE TO A FRIEND
69118 Sigma-Aldrich

1-Methyl-2-pyrrolidinone

puriss. p.a., ≥99.0% (GC)

Synonym: 1-Methyl-2-pyrrolidone, N-Methyl-2-pyrrolidone, NMP

  • CAS Number 872-50-4

  • Empirical Formula (Hill Notation) C5H9NO

  • Molecular Weight 99.13

  •  Beilstein Registry Number 106420

  •  EC Number 212-828-1

  •  MDL number MFCD00003193

  •  PubChem Substance ID 57652174

Purchase

Properties

Related Categories Analytical Reagents, Analytical Reagents for General Use, Analytical/Chromatography, I-N, Puriss p.a., Puriss p.a. More...
vapor density   3.4 (vs air)
vapor pressure   0.29 mmHg ( 20 °C)
  0.99 mmHg ( 40 °C)
InChI Key   SECXISVLQFMRJM-UHFFFAOYSA-N
grade   puriss. p.a.
assay   ≥99.0% (GC)
autoignition temp.   518 °F
expl. lim.   9.5 %
impurities   ≤0.1% water
refractive index   n20/D 1.47(lit.)
  n20/D 1.470
bp   202 °C(lit.)
  81-82 °C/10 mmHg(lit.)
mp   −24 °C(lit.)
solubility   water: miscible(lit.)
density   1.028 g/mL at 25 °C(lit.)
cation traces   Ca: ≤5 mg/kg
  Cd: ≤1 mg/kg
  Co: ≤1 mg/kg
  Cr: ≤1 mg/kg
  Cu: ≤1 mg/kg
  Fe: ≤1 mg/kg
  K: ≤20 mg/kg
  Mg: ≤1 mg/kg
  Mn: ≤1 mg/kg
  Na: ≤20 mg/kg
  Ni: ≤1 mg/kg
  Pb: ≤1 mg/kg
  Zn: ≤1 mg/kg

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
NA 1993 / PGIII
WGK Germany 
1
RTECS 
UY5790000
Flash Point(F) 
195.8 °F
Flash Point(C) 
91 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?