• USA Home
  • 69460 - N-Methyltrifluoroacetamide

EMAIL THIS PAGE TO A FRIEND
69460 Sigma-Aldrich

N-Methyltrifluoroacetamide

purum, ≥98.0% (GC)

  • CAS Number 815-06-5

  • Linear Formula CF3CONHCH3

  • Molecular Weight 127.07

  •  Beilstein Registry Number 1703392

  •  EC Number 212-417-7

  •  MDL number MFCD00009670

  •  PubChem Substance ID 24885928

Purchase

Properties

Related Categories Amides, Building Blocks, C2 to C7, Carbonyl Compounds, Chemical Synthesis,
InChI Key   IQNHBUQSOSYAJU-UHFFFAOYSA-N
grade   purum
assay   ≥98.0% (GC)
bp   156-157 °C(lit.)
mp   48-51 °C
  49-51 °C(lit.)

Description

Other Notes

Intermediate; starting material for the preparation of N-silylated derivatives; Gabriel-type reagent for the synthesis of secondary N-methylalkylamines

Same product — same quality — now Sigma-Aldrich brand

Price and Availability

Suggested Laboratory Gloves


Laboratory GlovesThis substance has been tested against several types of hand protection for CE compliance. Click below to find the recommended gloves for handling this product.



Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
UN1325 - class 4.1 - PG 2 - Flammable solids, organic, n.o.s., HI: all
WGK Germany 
3
Flash Point(F) 
165.2 °F
Flash Point(C) 
74 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?