• USA Home
  • 69482 - N-Methyl-N-(trimethylsilyl)trifluoroacetamide

EMAIL THIS PAGE TO A FRIEND
69482 Sigma-Aldrich

N-Methyl-N-(trimethylsilyl)trifluoroacetamide

purum, ≥97.0% (GC)

Synonym: N-Trimethylsilyl-N-methyl trifluoroacetamide, MSTFA

  • CAS Number 24589-78-4

  • Linear Formula CF3CON(CH3)Si(CH3)3

  • Molecular Weight 199.25

  •  Beilstein Registry Number 1941550

  •  EC Number 246-331-6

  •  MDL number MFCD00000411

Purchase

Properties

Related Categories Chemical Synthesis, Protecting and Derivatizing Reagents, Protection and Derivatization, Silicon-Based, Synthetic Reagents More...
vapor density   >1 (vs air)
vapor pressure   8.8 mmHg ( 27 °C)
InChI Key   MSPCIZMDDUQPGJ-UHFFFAOYSA-N
grade   purum
assay   ≥97.0% (GC)
refractive index   n20/D 1.379
  n20/D 1.38(lit.)
bp   130-132 °C(lit.)
density   1.075 g/mL at 25 °C(lit.)

Description

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
UN 1993C 3 / PGIII
WGK Germany 
3
RTECS 
AC9800000
Flash Point(F) 
78.8 °F
Flash Point(C) 
26 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

replaced by

Product #

Description

Add to Cart

394866 N-Methyl-N-(trimethylsilyl)trifluoroacetamide, synthesis grade

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?