• USA Home
  • 69555 - Nα-Methyl-L-tryptophan

EMAIL THIS PAGE TO A FRIEND
69555 Sigma-Aldrich

Nα-Methyl-L-tryptophan

purum, ≥98.0% (TLC)

  • CAS Number 526-31-8

  • Empirical Formula (Hill Notation) C12H14N2O2

  • Molecular Weight 218.25

  •  Beilstein Registry Number 86638

  •  EC Number 208-388-5

  •  MDL number MFCD00005645

  •  PubChem Substance ID 24885946

Purchase

Properties

Related Categories Amino Acids & Derivatives, Chemical Biology, Chemical Synthesis, Peptide Chemistry, Tryptophan More...
InChI Key   CZCIKBSVHDNIDH-NSHDSACASA-N
grade   purum
assay   ≥98.0% (TLC)
optical activity   [α]20/D +64±1°, c = 1% in 0.5 M NaOH
mp   ≥300 °C(lit.)

Description

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?