• USA Home
  • 69577 - α-Methyl-DL-tyrosine

EMAIL THIS PAGE TO A FRIEND
69577 Sigma-Aldrich

α-Methyl-DL-tyrosine

≥95% (NT)

Synonym: DL-2-Methyl-3-(4-hydroxyphenyl)alanine, AMPT

  • CAS Number 658-48-0

  • Linear Formula 4-(HO)C6H4CH2C(CH3)(NH2)CO2H

  • Molecular Weight 195.22

  •  Beilstein Registry Number 2938704

  •  EC Number 211-523-0

  •  MDL number MFCD00004187

  •  PubChem Substance ID 329761770

Purchase

Description

Biochem/physiol Actions

Tyrosine hydroxylase inhibitor.

Other Notes

Inhibitor (Ki 5 uM) of tyrosine hydroxylase

Price and Availability


Biomedical Applications
Safety & Documentation

Safety Information

WGK Germany 
3
RTECS 
YP2890000

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?