• USA Home
  • 69578 - α-Methyl-DL-tyrosine methyl ester hydrochloride

EMAIL THIS PAGE TO A FRIEND
69578 Sigma-Aldrich

α-Methyl-DL-tyrosine methyl ester hydrochloride

≥98.0% (TLC)

Synonym: α-MT methyl ester hydrochloride, 2-Methyl-4-hydroxy-DL-phenylalanine methyl ester hydrochloride, AMPT

  • CAS Number 7361-31-1

  • Linear Formula 4-(HO)C6H4CH2C(CH3)(NH2)CO2CH3 · HCl

  • Molecular Weight 245.70

  •  Beilstein Registry Number 5163031

  •  EC Number 230-900-0

  •  MDL number MFCD00012606

Purchase

Properties

Related Categories Amino Acids, Biochemicals and Reagents, Chemical Biology, Chemical Synthesis, I - Z,
InChI Key   OOVDEPZODSXAMU-UHFFFAOYSA-N
assay   ≥98.0% (TLC)
impurities   ≤2% water
mp   192 °C (dec.)(lit.)
solubility   H2O: soluble50 mg/mL, clear, faintly yellow
storage temp.   −20°C

Description

Biochem/physiol Actions

Tyrosine hydroxylase inhibitor.

Other Notes

Catecholamine depletor. Inhibitor of tyrosine hydroxylase; Prevention of nerve terminal damage in cerebral ischemia

Same product — same quality — now Sigma-Aldrich brand

Price and Availability

Safety & Documentation

Safety Information

WGK Germany 
3
RTECS 
YP2900000

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?