EMAIL THIS PAGE TO A FRIEND
69761 Sigma-Aldrich

(±)-Mevalonolactone

purum, ≥96.0% (T)

Synonym: (±)-β-Hydroxy-β-methyl-δ-valerolactone, (±)-3-Hydroxy-3-methyl δ-valerolactone, DL-Mevalolactone, DL-Mevalonic acid lactone

  • CAS Number 674-26-0

  • Empirical Formula (Hill Notation) C6H10O3

  • Molecular Weight 130.14

  •  Beilstein Registry Number 80960

  •  EC Number 211-615-0

  •  MDL number MFCD00006648

Purchase

Properties

Related Categories Allium cepa (Onion), Building Blocks, Carbonyl Compounds, Cell Biology, Chemical Synthesis,
InChI Key   JYVXNLLUYHCIIH-UHFFFAOYSA-N
grade   purum
assay   ≥96.0% (T)
impurities   salt, none detected
mp   24-27 °C
storage temp.   2-8°C

Description

Caution

can form fused mass at low temperatures

Other Notes

Lactone of mevalonic acid which is a very important connecting link in the biosynthesis of isoprenoids

Same product — same quality — now Sigma-Aldrich brand

Price and Availability


All labs need water
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
Flash Point(F) 
235.4 °F
Flash Point(C) 
113 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

replaced by

Product #

Description

Add to Cart

M4667 (±)-Mevalonolactone, ~97% (titration)

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?