• USA Home
  • 69765 - Micropipettes-Capillaries

EMAIL THIS PAGE TO A FRIEND
69765 Sigma-Aldrich

Micropipettes-Capillaries

Selectophore, borosilicate glass

Purchase

Properties

Related Categories Accessories for ISE, Analytical/Chromatography, Ion Sensor Materials More...
grade   Selectophore
O.D.   1.50 mm
diam.   1.20 mm

Description

Application

for the preparation of ion-selective microelectrodes

General description

Visit our Sensor Applications portal to learn more.

Legal Information

Selectophore is a trademark of Sigma-Aldrich Chemie GmbH

Price and Availability

Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?