• USA Home
  • 69810 - Molecular Weight Marker for proteins

EMAIL THIS PAGE TO A FRIEND
69810 Sigma-Aldrich

Molecular Weight Marker for proteins

for electrophoresis

Purchase

Properties

grade   for electrophoresis
mol wt   Mr 14200-67000
suitability   in accordance for electrophoresis test
storage temp.   2-8°C

Description

Components

SDS-PAGE-Mr:
α-lactalbumin from bovine milk: 14200; trypsin inhibitor from soybean: 20000; trypsinogen (PMSF-treated) from bovine pancreas: 24000; carbonic anhydrase from bovine erythrocytes: 29000; glyceraldehyde-3-phosphate dehydrogenase from rabbit muscle: 36000; albumin from chicken egg white: 45000; albumin from bovine serum: 67000

Price and Availability

Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?