• USA Home
  • 69824 - Mitomycin C from Streptomyces caespitosus

EMAIL THIS PAGE TO A FRIEND
69824 Sigma-Aldrich

Mitomycin C from Streptomyces caespitosus

  • CAS Number 50-07-7

  • Empirical Formula (Hill Notation) C15H18N4O5

  • Molecular Weight 334.33

  •  Beilstein Registry Number 7231816

  •  MDL number MFCD00078109

  •  PubChem Substance ID 329761995

Purchase

Description

Biochem/physiol Actions

Mode of Action: This product is an alkylating agent that specifically targets the guanine nucleoside sequence 5′-CpG-3′. It inhibits DNA synthesis by covalently reacting with DNA, forming crosslinks between complementary strands of DNA. This interaction prevents separation of complementary DNA strands, inhibiting DNA replication.

Antimicrobial Spectrum: Mitomycin C has strong antitumor activity, especially against Ehrlich ascites tumor cells, and strong bactericidal action against gram-positive and gram-negative bacteria.

Other Notes

Vial contains 2 mg mitomycin C and 48 mg NaCl (increases solubility).

Same product — same quality — now Sigma-Aldrich brand

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles

Articles

DNA Damage and Repair

Damage to cellular DNA is involved in mutagenesis and the development of cancer. The DNA in a human cell undergoes several thousand to a million damaging events per day, generated by both external (e...
Keywords: AGE, Alkylations, Apoptosis, Biochemistry, Cancer, Carcinogens, Catalysis, Clinical, DNA replication, Deaminations, Degradations, Eliminations, Environmental, Enzymology, Gene expression, Genetic, Infrared spectroscopy, Metabolites, Methylations, Mutagens, Oncology, Oxidations, Polymorphisms, Rearrangements, Recombination, Substitutions, Transcription, transformation

Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

replaced by

Product #

Description

Add to Cart

M4287 Mitomycin C from Streptomyces caespitosus, powder, BioReagent, suitable for cell culture

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?