• USA Home
  • 69831 - Molecular sieves, 3 Å

EMAIL THIS PAGE TO A FRIEND
69831 Sigma-Aldrich

Molecular sieves, 3 Å

rod

Synonym: Molecular Sieve UOP Type 3Å

Purchase

Properties

Related Categories Analytical Reagents, Analytical/Chromatography, Application Specific Reagents, Drying Agents
form   rod
size   1/8 in.
pore size   3 Å pore diameter

Description

Application

Molecular sieves may be employed in the enantioselective Friedel-Crafts alkylation of indoles with nitroalkenes. It may be employed for the drying of n-octane in a study.

General description

Molecular sieves are in the form of rods having pore diameter of 3Å. It participates in the synthesis of ecteinascidin 743 (a potential marine-derived antitumor agent) and vanillyl nonanoate (a model capsinoid compound).

Price and Availability


Laboratory Gloves
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?