• USA Home
  • 69833 - Molecular sieves, 3 Å

EMAIL THIS PAGE TO A FRIEND
69833 Sigma-Aldrich

Molecular sieves, 3 Å

powder

Synonym: Molecular Sieve UOP Type 3Å

Purchase

Properties

Related Categories Analytical Reagents, Analytical/Chromatography, Application Specific Reagents, Drying Agents
form   powder
pore size   3 Å pore diameter

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?