• USA Home
  • 69835 - Molecular sieves, 4 Å

EMAIL THIS PAGE TO A FRIEND
69835 Sigma-Aldrich

Molecular sieves, 4 Å

rod

Synonym: Sodium Alumino-Silicate

Purchase

Properties

Related Categories Analytical Reagents, Analytical/Chromatography, Application Specific Reagents, Drying Agents
form   rod
size   1/16 in.
pore size   4 Å pore diameter

Description

Application

Molecules with an effective diameter of 4 Å, including H2O, H2S, CO2, SO2, C2H4 and C3H6 are adsorbed.

Molecular sieves, 4 Å may be employed in the synthesis of methyl 8-methylnon-6-enoate and methyl 8-methylnonanoate.

General description

Molecular sieves are in the form of the rods having pore diameter of 4Å. It participates in the synthesis of ecteinascidin 743 (a potential marine-derived antitumor agent).

Price and Availability


Laboratory Gloves
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?