• USA Home
  • 69846 - Molybdenum(II) acetate dimer

EMAIL THIS PAGE TO A FRIEND
69846 Sigma-Aldrich

Molybdenum(II) acetate dimer

purum, ≥98.0% (CH)

Synonym: Di-molybdenum tetraacetate

Purchase

Properties

Related Categories Catalysis and Inorganic Chemistry, Chemical Synthesis, Molybdenum More...
InChI Key   DOOLFANBWPPEGQ-UHFFFAOYSA-J
grade   purum
assay   ≥98.0% (CH)
shipped in   wet ice

Description

Other Notes

Complexing agent used in the determination of absolute configuration of vicinal glycols by circular dichroism

Price and Availability


Laboratory Gloves
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

232076 Molybdenum(II) acetate dimer, 98%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?