• USA Home
  • 69902 - 4-Morpholinecarbonyl chloride

EMAIL THIS PAGE TO A FRIEND
69902 Sigma-Aldrich

4-Morpholinecarbonyl chloride

purum, ≥98.0% (T)

  • CAS Number 15159-40-7

  • Empirical Formula (Hill Notation) C5H8ClNO2

  • Molecular Weight 149.58

  •  Beilstein Registry Number 116257

  •  EC Number 239-213-0

  •  MDL number MFCD00037053

  •  PubChem Substance ID 329762102

Purchase

Properties

Related Categories Building Blocks, C4 to C10, Chemical Synthesis, Heterocyclic Building Blocks, Morpholines/Thiomorpholines More...
InChI Key   XMWFMEYDRNJSOO-UHFFFAOYSA-N
grade   purum
assay   ≥98.0% (T)
refractive index   n20/D 1.498(lit.)
  n20/D 1.500
bp   137-138 °C/33 mmHg(lit.)
density   1.282 g/mL at 25 °C(lit.)

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Warning
Hazard statements 
Precautionary statements 
Supplemental Hazard Statements 
Reacts violently with water.
RIDADR 
UN 3265 8 / PGII
WGK Germany 
3
Flash Point(F) 
235.4 °F
Flash Point(C) 
113 °C
Protocols & Articles
Related Products

replaced by

Product #

Description

Add to Cart

348295 4-Morpholinecarbonyl chloride, 98%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?